Human Recombinant Galectin 14 (from Cells)
Fournisseur: ProSci Inc.
91-644
PRSI91-644EA 789 EUR InStock
PRSI91-644
Human Recombinant Galectin 14 (from Cells)
Protéines et Peptides
Galectin-14 is a member of the Galectin family of carbohydrate binding proteins. The members of Galectin family contain one or two carbohydrate recognition domains, which can bind beta -Galactoside. LGALS14 is expressed intracellularly in placenta and eosinophils, and is released by eosinophils following allergen stimulation. LGALS14 may be involved in the development of allergic inflammation. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité