Human recombinant PRPS2 (from cells)
Fournisseur: ProSci Inc.
91-645
PRSI91-645EA 654 EUR InStock
PRSI91-645
Human recombinant PRPS2 (from cells)
Protéines et Peptides
Ribose-Phosphate Pyrophosphokinase 2 (PRPS2) is a phosphoribosyl pyrophosphate synthetase that belongs to the ribose-phosphate pyrophosphokinase family. PRPS2 is a homodimer. The active form is probably an hexamer composed of three homodimers. PRPS2 catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis. PRPS2 catalyzes the synthesis of 5-phosphoribosyl 1-pyrophosphate from ATP and D-ribose 5-phosphate. In addition, PRPS2 plays a central role in the synthesis of purines and pyrimidines.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité