Human recombinant BMPG (from Cells)
Fournisseur: ProSci Inc.
91-647
PRSI91-647EA 789 EUR InStock
PRSI91-647
Human recombinant BMPG (from Cells)
Protéines et Peptides
Bone Marrow Proteoglycan (BMPG) is a secreted protein that contains one C-type lectin domain. BMPG is the predominant constituent of the crystalline core of the eosinophil granule. BMPG is highly expressed in placenta and pregnancy serum. BMPG may be involved in antiparasitic defense mechanisms as a cytotoxin and helminthotoxin. BMPG induces non-cytolytic histamine release from human basophils. In addition, BMPG also participated in antiparasitic defense mechanisms and immune hypersensitivity reactions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité