Human Recombinant HATL5 (from Cells)
Fournisseur: ProSci Inc.
91-649
PRSI91-649EA 884 EUR InStock
PRSI91-649
Human Recombinant HATL5 (from Cells)
Protéines et Peptides
Transmembrane Protease Serine 11B (TMPRSS11B) is a single-pass type II membrane protein member of the peptidase S1 family. TMPRSS11B contains one peptidase S1 domain and one SEA domain. TMPRSS11B is a serine protease that may play some biological role in the host defense system on the mucous membrane independently of or in cooperation with other substances in airway mucous or bronchial secretions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité