Human Recombinant GPD1 (from Cells)
Fournisseur: ProSci Inc.
91-655
PRSI91-655EA 884 EUR InStock
PRSI91-655
Human Recombinant GPD1 (from Cells)
Protéines et Peptides
Glycerol-3-Phosphate Dehydrogenase [NAD(+)], Cytoplasmic (GPDH-C) belongs to the NAD-Dependent Glycerol-3-Phosphate Dehydrogenase family. GPDH-C plays a critical role in carbohydrate and lipid metabolism by catalyzing the reversible conversion of Dihydroxyacetone Phosphate (DHAP) and reducing Nicotine Adenine Dinucleotide (NADH) to Glycerol-3-Phosphate (G3P) and NAD+. GPDH-C is inhibited by zinc ions and sulfate. Mutations in this gene are a cause of transient infantile hypertriglyceridemia. GPDH-C is unlike Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH); they have different substrates.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité