Human recombinant VIP36 (from cells)
Fournisseur: ProSci Inc.
91-659
PRSI91-659EA 584 EUR InStock
PRSI91-659
Human recombinant VIP36 (from cells)
Protéines et Peptides
Vesicular integral-membrane protein VIP36 is also known as Glycoprotein GP36b, Lectin mannose-binding 2, Vesicular integral-membrane protein 36, LMAN2 and C5orf8. LMAN2 is widely expressed and contains one L-type lectin-like domain. LMAN2 binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control. LMAN2 plays a role as an intracellular lectin in the early secretory pathway. LMAN2 interacts with N-acetyl-D-galactosamine and high-mannose type glycans and may also bind to O-linked glycans. LMAN2 is also involved in the transport and sorting of glycoproteins carrying high mannose-type glycans.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité