Human Recombinant 15-PGDH (from Cells)
Fournisseur: ProSci Inc.
91-661
PRSI91-661EA 884 EUR InStock
PRSI91-661
Human Recombinant 15-PGDH (from Cells)
Protéines et Peptides
15-hydroxyprostaglandin dehydrogenase [NAD(+)], also known as Prostaglandin dehydrogenase 1, 15-PGDH, HPGD and PGDH1, belongs to the short-chain dehydrogenases/reductases (SDR) family. HPGD localizes to the cytoplasm and can be found in colon epithelium, existing as a homodimer. HPGD catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. HPGD is down-regulated by cortisol, dexamethasone and betamethasone, up-regulated by TGFB1. HPGD inhibits in vivo proliferation of colon cancer cells. HPGD is the key enzyme for the inactivation of prostaglandins, and thus regulates processes such as inflammation or proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité