Human recombinant UBE2B (from cells)
Fournisseur: ProSci Inc.
91-665
PRSI91-665EA 654 EUR InStock
PRSI91-665
Human recombinant UBE2B (from cells)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 B (UBE2B) is a member of the ubiquitin-conjugating enzyme family. UBE2B accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. It is shown that UBE2B interacts with RAD18, UBR2, and WAC. UBE2B is required for post-replicative DNA damage repair. Additional, UBE2B plays a role in sepsis-induced muscle protein proteolysis and cancer-induced cachexia.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité