Human recombinant GMP Reductase 1 (from cells)
Fournisseur: ProSci Inc.
91-666
PRSI91-666EA 654 EUR InStock
PRSI91-666
Human recombinant GMP Reductase 1 (from cells)
Enzymes
GMP Reductase 1 (GMPR) is a member of the IMPDH/GMPR family. GMPR exists as a homotetramer and catalyses the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides. GMP reductase gene expression may be regulated by MITF. At least two different alleles are known.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité