Human recombinant fumarylacetoacetase (from cells)
Fournisseur: ProSci Inc.
91-668
PRSI91-668EA 761 EUR InStock
PRSI91-668
Human recombinant fumarylacetoacetase (from cells)
Enzymes
Fumarylacetoacetase belongs to the FAH family. Fumarylacetoacetase is primary expressed in liver and kidney. It exists as a homodimer and catalyses the hydrolysis of 4-fumarylacetoacetate into fumarate and acetoacetate. Defects in Fumarylacetoacetase cause tyrosinemia type 1, which is congenital metabolism defect characterised by elevated levels of tyrosine in the blood and urine, and hepatorenal manifestations. Typical features include renal tubular injury, self-mutilation, hepatic necrosis, episodic weakness, and seizures.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité