Human recombinant trefoil factor 2 (from cells)
Fournisseur: ProSci Inc.
91-669
PRSI91-669EA 654 EUR InStock
PRSI91-669
Human recombinant trefoil factor 2 (from cells)
Protéines et Peptides
Trefoil Factor 2 (TFF2) is a member of the trefoil family and contains two P-type (trefoil) domains. Members of this family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. TFF2 is a secreted protein and specifically expressed in the stomach. TFF2 inhibits gastrointestinal motility and gastric acid secretion. TFF2 could function as a structural component of gastric mucus, possibly by stabilizing glycoproteins in the mucus gel through interactions with carbohydrate side chains.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité