Human recombinant proteoglycan 3 (from cells)
Fournisseur: ProSci Inc.
91-673
PRSI91-673EA 654 EUR InStock
PRSI91-673
Human recombinant proteoglycan 3 (from cells)
Protéines et Peptides
Proteoglycan 3, also known as Eosinophil major basic protein homolog, Prepro-major basic protein homolog, PRG3 and MBPH, contains one C-type lectin domain. Proteoglycans are a major component of the animal extracellular matrix. PRG3 localizes to the eosinophil secondary granule and is expressed in bone marrow, not detected in placenta. PRG3 has similar cytotoxic and cytostimulatory activities to PRG2/MBP. In vitro, PRG3 can stimulate neutrophil superoxide production and IL8 release, histamine and leukotriene C4 release from basophils.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité