Human recombinant trans-2-Enoyl-CoA Reductase (from cells)
Fournisseur: ProSci Inc.
91-680
PRSI91-680EA 761 EUR InStock
PRSI91-680
Human recombinant trans-2-Enoyl-CoA Reductase (from cells)
Enzymes
Trans-2-Enoyl-CoA Reductase Mitochondrial (MECR) belongs to the zinc-containing alcohol dehydrogenase family. MECR localises to the mitochondrion. It is highly expressed in skeletal and heart muscle and expressed at lower levels in the placenta, liver, kidney and pancreas, with weakly or no expression in the lung. MECR exists as a homodimer, which catalyses the reduction of trans-2-enoyl-CoA to acyl-CoA with chain length from C6 to C16 in an NADPH-dependent manner with preference to medium chain length substrate. MECR may take part in the mitochondrial synthesis of fatty acids.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité