Human recombinant ER alpha-1,2-Mannosidase (from cells)
Fournisseur: ProSci Inc.
91-681
PRSI91-681EA 584 EUR InStock
PRSI91-681
Human recombinant ER alpha-1,2-Mannosidase (from cells)
Enzymes
Endoplasmic Reticulum Mannosyl-Oligosaccharide 1,2- alpha-Mannosidase (MAN1B1) belongs to the glycosyl hydrolase 47 family. MAB1B1 is a single-pass type II membrane protein and widely expressed in many tissues. MAB1B1 is involved in glycoprotein quality control targeting of misfolded glycoproteins for degradation. MAB1B1 can be inhibited by both 1-deoxymannojirimycin (dMNJ) and kifunensine. Defects in MAN1B1 are the cause of mental retardation autosomal recessive type 15 (MRT15). Mental retardation is characterised by significantly below average general intellectual functioning, it is also associated with impairments in adaptative behavior and manifested during the developmental period.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité