Human recombinant CEACAM7 (from cells)
Fournisseur: ProSci Inc.
91-684
PRSI91-684EA 852 EUR InStock
PRSI91-684
Human recombinant CEACAM7 (from cells)
Protéines et Peptides
Carcinoembryonic Antigen-Related Cell Adhesion Molecule 7 (CEACAM7) is a member of the immunoglobulin superfamily A and CEA family. CEACAM7 localizes to the cell membrane and contains one Ig-like C2-type domain and one Ig-like V-type domain. The expression of CEACAM7 is significantly decreased in rectal cancer. Differences in CEACAM7 expression levels between long-term survivors and those with recurrent disease introduce a potential tumor marker to define a subset of patients who benefit most from adjuvant therapy.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité