Human recombinant SHMT1 (from cells)
Fournisseur: ProSci Inc.
91-687
PRSI91-687EA 852 EUR InStock
PRSI91-687
Human recombinant SHMT1 (from cells)
Protéines et Peptides
Serine Hydroxymethyltransferase Cytosolic (SHMT1) is a member of the SHMT family. SHMT1 is a cytoplasmic protein and exists as a homotetramer. SHMT1 catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon unit for the synthesis of methionine, thymidylate, and purines in the cytoplasm. A reduction in SHMT1 levels would result in less glycine that could affect the nervous system by acting as an agonist to the NMDA receptor and this could be a mechanism behind Smith-Magenis syndrome.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité