Human recombinant neuropeptide Y (from cells)
Fournisseur: ProSci Inc.
91-688
PRSI91-688EA 654 EUR InStock
PRSI91-688
Human recombinant neuropeptide Y (from cells)
Protéines et Peptides
Pro-Neuropeptide Y (NPY) is a member of the NPY family. NPY is a secreted protein and is one of the most abundant peptides in the nervous system. It also can be found in some chromaffin cells of the adrenal medulla. NPY can be cleaved into Neuropeptide Y and C-flanking peptide of NPY chain, which regulates energy usage, and it is involved in learning, memory processing, and epilepsy. NPY is implicated in the control of feeding and in secretion of gonadotrophin-release hormone. In addition, NPY increases the proportion of energy stored as fat and blocks nociceptive signals to the brain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité