Human Recombinant Golgi Membrane 1 (from Cells)
Fournisseur: ProSci Inc.
91-689
PRSI91-689EA 654 EUR InStock
PRSI91-689
Human Recombinant Golgi Membrane 1 (from Cells)
Protéines et Peptides
Golgi Membrane Protein 1 (GOLM1) belongs to the GOLM1/CASC4 family, GOLM1 is a single-pass type II membrane protein and can be found in many tissues . GOLM1 is overexpressed in prostate cancer and lung adenocarcinoma tissue. GOLM1 can be up-regulated in response to viral infection. GOLM1 is induced by the E1A adenoviral protein. GOLM1 plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité