Human recombinant CaM Kinase I (from Cells)
Fournisseur: ProSci Inc.
91-691
PRSI91-691EA 584 EUR InStock
PRSI91-691
Human recombinant CaM Kinase I (from Cells)
Enzymes
Calcium/Calmodulin-Dependent Protein Kinase Type 1 (CAMK1) belongs to the protein kinase superfamily, CAMK Ser/Thr protein kinase family, and CaMK subfamily. CAMK1 contains one protein kinase domain and widely expressed. CAMK1 is phosphorylated by CaMKK1 and CaMKK2 on Thr-177. CAMK1 regulates transcription activators activity, cell cycle, hormone production, cell differentiation, actin filament organisation, and neurite outgrowth. CAMK1 plays a role in K+ and ANG2-mediated regulation of the aldosterone synthase (CYP11B2) to produce aldosterone in the adrenal cortex.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité