Human recombinant Collagen alpha-1(VIII) chain (from cells)
Fournisseur: ProSci Inc.
91-692
PRSI91-692EA 761 EUR InStock
PRSI91-692
Human recombinant Collagen alpha-1(VIII) chain (from cells)
Protéines et Peptides
Collagen alpha-1(VIII) chain, also known as endothelial collagen, C3orf7 and COL8A1, can be cleaved into vastatin chain. COL8A1 is a short chain collagen and a major component of the basement membrane of the corneal endothelium. COL8A1 forms homotrimers, or heterotrimers in association with alpha 2(VIII) type collagens. Four homotrimers can form a tetrhedron stabilized by central interacting C-terminal NC1 trimers. COL8A1 contains one C1q domain and is primarily expressed in the subendothelium of large blood vessels. The expression level can be up-regulated during vascular injury, in atherosclerosis and in diabetes. COL8A1 may have a role in the maintenance of vessel wall integrity and structure, in particular in atherogenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité