Human recombinant IL-22 binding (from cells)
Fournisseur: ProSci Inc.
91-699
PRSI91-699EA 654 EUR InStock
PRSI91-699
Human recombinant IL-22 binding (from cells)
Protéines et Peptides
Interleukin-22 Receptor Subunit alpha-2 (IL22RA2) belongs to the type II cytokine receptor family. IL22RA2 is a secreted protein and contains three fibronectin type-III domains. IL22RA2 is widely expressed in many tissues. IL22RA2 functions as an IL22 antagonist and may be important in the regulation of inflammatory response. Three alternatively spliced transcript variants encoding distinct isoforms have been described.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité