Human recombinant VDB (from cells)
Fournisseur: ProSci Inc.
91-700
PRSI91-700EA 584 EUR InStock
PRSI91-700
Human recombinant VDB (from cells)
Protéines et Peptides
Vitamin D-Binding Protein (DBP) is a member of the ALB/AFP/VDB family. DBP is a secreted protein and contains three albumin domains. The primary structure contains 28 cysteine residues forming multiple disulfide bonds. DBP acts as a multifunctional protein found in plasma, ascitic fluid, cerebrospinal fluid, and urine and on the surface of many cell types. DBP binds to vitamin D and its plasma metabolites and transports them to target tissues. DBP associates with membrane-bound immunoglobulin on the surface of B-lymphocytes and with IgG Fc receptor on the membranes of T-lymphocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité