Human recombinant Pyruvate Kinase (from cells)
Fournisseur: ProSci Inc.
91-701
PRSI91-701EA 584 EUR InStock
PRSI91-701
Human recombinant Pyruvate Kinase (from cells)
Enzymes
Pyruvate Kinase Isozymes R/L (PKLR) belongs to the pyruvate kinase family, There are 4 isozymes of pyruvate kinase in mammals: L, R, M1 and M2. L type is major isozyme in the liver; R is found in red cells; M1 is the main form in muscle, heart and brain; M2 is found in early fetal tissues. PKLR exists as a homotetramer and catalyses the production of phosphoenolpyruvate from pyruvate and ATP. Defects in PKLR are also the cause of pyruvate kinase deficiency of red cells, which is a frequent cause of hereditary non-spherocytic hemolytic anemia.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité