Human recombinant FLRT3 (from cells)
Fournisseur: ProSci Inc.
91-706
PRSI91-706EA 654 EUR InStock
PRSI91-706
Human recombinant FLRT3 (from cells)
Protéines et Peptides
Leucine-Rich Repeat Transmembrane Protein FLRT3 (FLRT3) is a member of the fibronectin leucine rich transmembrane protein (FLRT) family. Proteins in this family play an role in cell adhesion and/or receptor signalling. FLRT3 is a single-pass type I membrane protein and contains one fibronectin type-III domain, ten LRR (leucine-rich) repeats, one LRRCT domain, and one LRRNT domain. FLRT3 may have a function in cell adhesion and/or receptor signaling. FLRT3 may regulate cellular adhesion between the epithelial apical ridge and the underlying mesenchyme and in establishing the dorso-ventral position of the ridge.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité