Human recombinant Aldo-Keto Reductase 1C3 (from cells)
Fournisseur: ProSci Inc.
91-710
PRSI91-710EA 678 EUR InStock
PRSI91-710
Human recombinant Aldo-Keto Reductase 1C3 (from cells)
Enzymes
AKR1C3, is an enzyme which belongs to the aldo/keto reductase family. It is expressed in many tissues including adrenal gland, brain, kidney, liver, lung, mammary gland, placenta, small intestine, colon, spleen, prostate and testis. AKR1C3 catalyses the conversion of aldehydes and ketones to alcohols. It catalyses the reduction of prostaglandin (PG) D2, PGH2 and phenanthrenequinone (PQ) and the oxidation of 9-alpha,11-beta-PGF2 to PGD2,which functions as a bi-directional 3-alpha-, 17-beta- and 20-alpha HSD. It can interconvert active androgens, estrogens and progestins with their cognate inactive metabolites.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité