Human recombinant neu-1 (from cells)
Fournisseur: ProSci Inc.
91-715
PRSI91-715EA 884 EUR InStock
PRSI91-715
Human recombinant neu-1 (from cells)
Protéines et Peptides
Sialidase-1 belongs to the N-acetyl-a neuraminidase family. Sialidase-1 is expressed in many tissues; it is highly expressed in the pancreas, and weakly expressed in the brain. Sialidase-1 is a lysosomal enzyme, which cleaves terminal sialic acid residues from substrates such as glycoproteins and glycolipids. Deficiencies in the human enzyme Sialidase-1 leads to sialidosis, a rare lysosomal storage disease. Sialidase-1 has been shown to interact with Cathepsin A (protective protein), beta -galactosidase and N-acetylgalactosamine-6-sulfate sulfatase in a multienzyme complex.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité