Human recombinant Apolipoprotein A-IV (from cells)
Fournisseur: ProSci Inc.
91-720
PRSI91-720EA 852 EUR InStock
PRSI91-720
Human recombinant Apolipoprotein A-IV (from cells)
Protéines et Peptides
Apolipoprotein A4 (APOA4) is a secreted protein that belongs to the apolipoprotein A1/A4/E family. Apoa-IV is a major component of HDL and chylomicrons. APOA4 is secreted into circulation on the surface of newly synthesized chylomicron particles. APOA4 play a role in the regulation of appetite and satiety in rodent models. APOA4 involved in chylomicrons and VLDL secretion and catabolism and required for efficient activation of lipoprotein lipase by ApoC-II. In addition, APOA4 is a potent activator of lecithin-cholesterol acyltransferase in vitro.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité