Human recombinant PDL2 (from cells)
Fournisseur: ProSci Inc.
91-721
PRSI91-721EA 852 EUR InStock
PRSI91-721
Human recombinant PDL2 (from cells)
Protéines et Peptides
Programmed Cell Death 1 Ligand 2 (PDCD1LG2) is a member of the BTN/MOG family. PDCD1LG2 contains one Ig-like C2-type domain and one Ig-like V-type domain. PDCD1LG2 is highly expressed in the heart, placenta, pancreas, lung and liver; it is weakly expressed in the spleen, lymph nodes, and thymus. PDCD1LG2 is involved in the costimulatory signal, essential for T-cell proliferation and IFNG production in a PDCD1-independent manner. PDCD1LG2 interaction with PDCD1 inhibits T-cell proliferation by blocking cell cycle progression and cytokine production.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité