Human recombinant Cadherin-8 (from Cells)
Fournisseur: ProSci Inc.
91-722
PRSI91-722EA 852 EUR InStock
PRSI91-722
Human recombinant Cadherin-8 (from Cells)
Protéines et Peptides
Cadherin-8 (CDH8) is a type II classical cadherin from the cadherin superfamily. Member of the Cadherin superfamily are integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small highly conserved C-terminal cytoplasmic domain. Cadherins are calcium dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells. The extracellular domain of CDH8 contains five cadherin domains. CDH8 is expressed in brain and is putatively involved in synaptic adhesion, axon outgrowth and guidance.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité