Human recombinant uroplakin 2 (from cells)
Fournisseur: ProSci Inc.
91-723
PRSI91-723EA 852 EUR InStock
PRSI91-723
Human recombinant uroplakin 2 (from cells)
Protéines et Peptides
Uroplakin-2 is a single-pass type I membrane protein that belongs to the uroplakin-2 family. Uroplakin-2 is a component of the asymmetric unit membrane (AUM) and expressed in the ureter, a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. Uroplakin-2 forms heterodimer with UPK1A that is necessary for exiting out of the endoplasmic reticulum (ER). Uroplakin-2 may play an important role in regulating the assembly of the AUM. AUM is believed to strengthen the urothelium by preventing cell rupture during bladder distention.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité