Human recombinant Follitropin Subunit beta (from cells)
Fournisseur: ProSci Inc.
91-725
PRSI91-725EA 852 EUR InStock
PRSI91-725
Human recombinant Follitropin Subunit beta (from cells)
Protéines et Peptides
Follitropin Subunit beta (FSHB) is a secreted protein that belongs to the glycoprotein hormones subunit beta family. The pituitary glycoprotein hormone family includes follicle-stimulating hormone, luteinizing hormone, chorionic gonadotropin, and thyroid-stimulating hormone. FSHB exists in a heterodimer of a common alpha chain and an unique beta chain that confers biological specificity to thyrotropin, lutropin, follitropin, and gonadotropin. FSHB stimulates development of follicle and spermatogenesis in the reproductive organs. Defects in FSHB are a cause of isolated follicle-stimulating hormone deficiency (IFSHD).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité