Human recombinant sulphamidase (from cells)
Fournisseur: ProSci Inc.
91-726
PRSI91-726EA 789 EUR InStock
PRSI91-726
Human recombinant sulphamidase (from cells)
Enzymes
N-Sulphoglucosamine Sulphohydrolase (SGSH) is an important member of the sulphatase family which is involved in the degradation of heparin sulphate. SGSH binds one calcium ion per subunit as a cofactor. SGSH catalyses N-sulfo-D-glucosamine and H₂O to D-glucosamine and sulphate. SGSH deficiency is result in mucopolysaccharidosis type 3A (MPS3A), a recessive lysosomal storage disease characterised by neurological dysfunction but relatively mild somatic manifestations.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité