Human recombinant Mucin 15 (from cells)
Fournisseur: ProSci Inc.
91-727
PRSI91-727EA 852 EUR InStock
PRSI91-727
Human recombinant Mucin 15 (from cells)
Protéines et Peptides
Mucin-15 is a single-pass type I membrane protein member of the Mucin family. Mucins are a family of high molecular weight, heavily glycosylated proteins (glycoconjugates) produced by epithelial tissues in most metazoans. A key characteristic of Mucins is their ability to form gels. Therefore they are a key component in most gel-like secretions, serving functions from lubrication to cell signalling to forming chemical barriers. Mucin-15 is expressed in many tissues. Mucin-15 is highly glycosylated (N- and O-linked carbohydrates). Mucin-15 may play a role in the cell adhesion to the extracellular matrix.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité