Human recombinant IL9 (from cells)
Fournisseur: ProSci Inc.
91-728
PRSI91-728EA 852 EUR InStock
PRSI91-728
Human recombinant IL9 (from cells)
Protéines et Peptides
Interleukin-9 (IL-9) is a secreted protein that belongs to the IL-7/IL-9 family. IL-9 supports IL-2 independent and IL-4 independent growth of helper T-cells. IL-9 stimulates cell proliferation and prevents apoptosis. It functions through the IL-9 receptor (IL-9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. IL-9 has been identified as a candidate gene for asthma. IL-9 is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité