Human recombinant sialate O-Acetylesterase (from cells)
Fournisseur: ProSci Inc.
91-729
PRSI91-729EA 884 EUR InStock
PRSI91-729
Human recombinant sialate O-Acetylesterase (from cells)
Enzymes
Sialate O-Acetylesterase (SIAE) belongs to the family of hydrolases, specifically those acting on carboxylic ester bonds. SIAE is widely expressed with high expression levels in the testis, prostate, and colon. SIAE catalyses N-acetyl-O-acetylneuraminate and H2O to N-acetylneuraminate and acetate. SIAE removes O-acetyl ester groups from position 9 of the parent sialic acid, N-acetylneuraminic acid. SIAE down-regulates B lymphocyte antigen receptor signaling (involving CD22), and is required for immunological tolerance. Loss of function mutations in SIAE are much more frequently found in humans with autoimmune diseases especially rheumatoid arthritis and type 1 diabetes.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité