Human Recombinant CD116 (from Cells)
Fournisseur: ProSci Inc.
91-733
PRSI91-733EA 588 EUR InStock
PRSI91-733
Human Recombinant CD116 (from Cells)
Protéines et Peptides
Granulocyte-Macrophage Colony-Stimulating Factor Receptor Subunit alpha (CSF2RA) is a single-pass type I membrane protein which belongs to the type I cytokine receptor family of Type 5 subfamily. The CSF2RA gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes with some of the isoforms being membrane-bound and others being soluble. CSF2RA is a low affinity receptor for granulocyte-macrophage colony-stimulating factor. CSF2RA transduces a signal that results in the proliferation, differentiation, and functional activation of hematopoietic cells. Defects in CSF2RA are the cause of pulmonary surfactant metabolism dysfunction type 4 (SMDP4).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité