Human recombinant IL28B (from cells)
Fournisseur: ProSci Inc.
91-735
PRSI91-735EA 761 EUR InStock
PRSI91-735
Human recombinant IL28B (from cells)
Protéines et Peptides
Interleukin-28B, also known as Cytokine Zcyto22, Interferon lambda-3, Interferon lambda-4, IFNL3, IFNL4, ZCYTO22 and IL28B, is a secreted cytokine which belongs to the IL-28/IL-29 family. IL-28 has also been shown to play a role in the adaptive immune response. IL28B has immunomodulatory activity and up-regulates MHC class I antigen expression. IL28B displays potent antiviral activity and antitumor activity. In addition, IL28B is a ligand for the heterodimeric class II cytokine receptor composed of IL10RB and IL28RA. The ligand/receptor complex seems to signal through the Jak-STAT pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité