Human recombinant CD79B (from cells)
Fournisseur: ProSci Inc.
91-738
PRSI91-738EA 852 EUR InStock
PRSI91-738
Human recombinant CD79B (from cells)
Protéines et Peptides
CD79B is a single-pass type I membrane protein. CD79B contains one Ig-like V-type domain and one ITAM domain. CD79B is required in cooperation with CD79A for initiation of the signal transduction cascade activated by the B-cell antigen receptor complex (BCR), which leads to internalization of the complex, trafficking to late endosomes and antigen presentation. CD79B enhances phosphorylation of CD79A, possibly by recruiting kinases that phosphorylate CD79A or by recruiting proteins that bind to CD79A and protect it from dephosphorylation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité