Human recombinant tripeptidyl-peptidase I (from cells)
Fournisseur: ProSci Inc.
91-745
PRSI91-745EA 884 EUR InStock
PRSI91-745
Human recombinant tripeptidyl-peptidase I (from cells)
Enzymes
Tripeptidyl-Peptidase 1 (TPP1) belongs to the peptidase S53 family. TPP1 is detected in all tissues examined with highest levels in heart and placenta and relatively similar levels in other tissues. TPP1 is lysosomal serine protease with tripeptidyl-peptidase I activity. TPP1 may act as a non-specific lysosomal peptidase which generates tripeptides from the breakdown products produced by lysosomal proteinases. TPP1 requires substrates with an unsubstituted N-terminus. TPP1 mutations have also been shown to cause neuronal ceroid lipofuscinosis type 2 (CLN2).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité