Human recombinant Elafin (from cells)
Fournisseur: ProSci Inc.
91-748
PRSI91-748EA 884 EUR InStock
PRSI91-748
Human recombinant Elafin (from cells)
Protéines et Peptides
Elafin is a secreted protein. Elafin consists of two domains: the transglutaminase substrate domain (cementoin moiety) and the elastase inhibitor domain. The transglutaminase substrate domain serves as an anchor to localize Elafin covalently to specific sites on extracellular matrix proteins. It is shown that Elafin can be used as a biomarker for graft versus host disease of the skin. Elafin is expressed in the skin during wound healing through activation of the epidermal growth factor receptor. Elafin may prevent elastase-mediated tissue proteolysis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité