Human recombinant reticulocalbin 3 (from cells)
Fournisseur: ProSci Inc.
91-752
PRSI91-752EA 584 EUR InStock
PRSI91-752
Human recombinant reticulocalbin 3 (from cells)
Protéines et Peptides
Reticulocalbin-3 is a member of the CREC family that is involved in the secretory pathway. Members of the CREC family consist of a number of multiple EF-hand domains. Reticulocalbin-3 localizes to the endoplasmic reticulum lumen. The bioinformaticanalysis of Reticulocalbin-3 reveal that it is a putative Ca2+-binding protein. In addition, it has been shown that autoactivation and secretion of PACE4 was increased upon co-expression with Reticulocalbin-3. The proPACE4-RCN-3 complex is plays an important role in the biosynthesis of PACE4.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité