Human recombinant Cadherin-11 (from Cells)
Fournisseur: ProSci Inc.
91-753
PRSI91-753EA 654 EUR InStock
PRSI91-753
Human recombinant Cadherin-11 (from Cells)
Protéines et Peptides
Cadherin-11 is a type II classical cadherin member of the cadherin superfamily of integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Cadherins interact with themselves in a homophilic manner in connecting cells, and thus contribute to the sorting of heterogeneous cell types. Cadherin-11 contains five cadherin domains and is mainly expressed in the brain. Mature cadherin proteins consists of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small highly conserved C-terminal cytoplasmic domain. It is shown that Cadherin-11 is a viable molecular target for therapeutic intervention in Glioblastoma multiforme.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité