Human recombinant LILRA2 (from cells)
Fournisseur: ProSci Inc.
91-754
PRSI91-754EA 654 EUR InStock
PRSI91-754
Human recombinant LILRA2 (from cells)
Protéines et Peptides
Leukocyte Immunoglobulin-Like Receptor Subfamily A Member 2 (LILRA2) is a single-pass type I membrane protein. LILRA2 is expressed predominantly on monocytes and B cells, and at lower levels on dendritic cells and natural killer cells. LILRA2 contains four Ig-like C2-type domains, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades. LILRA2 does not bind class I MHC antigens.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité