Human recombinant serpin A10 (from cells)
Fournisseur: ProSci Inc.
91-759
PRSI91-759EA 654 EUR InStock
PRSI91-759
Human recombinant serpin A10 (from cells)
Protéines et Peptides
Serpin A10 is a secreted protein that belongs to the serpin family. It is predominantly expressed in the liver and secreted in the plasma. Its phosphorylation sites are present in the extracelllular medium. It inhibits factors Xa and XIa of the coagulation cascade in the presence of protein Z, calcium, and phospholipid. Mutations in SERPINA10 are associated with venous thrombosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité