Human recombinant Collectin 11 (from cells)
Fournisseur: ProSci Inc.
91-761
PRSI91-761EA 525 EUR InStock
PRSI91-761
Human recombinant Collectin 11 (from cells)
Protéines et Peptides
Collectin-11 is a secreted protein that belongs to the COLEC10/COLEC11 family. Collectin-11 contains one C-type lectin domain and one collagen-like domain. Collectins play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. Collectin-11 binds to various sugars including fucose and mannose, but does not bind to glucose, N-acetylglucosamine and N-acetylgalactosamine. It has a higher affinity for fucose compared to mannose. Collectin-11 binds lipopolysaccharides (LPS). It also involved in fundamental development serving as a guidance cue for neural crest cell migration. Defects in Collectin-11 are the cause of 3MC syndrome type 2 (3MC2).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité