Human recombinant ER Resident 27 (from cells)
Fournisseur: ProSci Inc.
91-767
PRSI91-767EA 584 EUR InStock
PRSI91-767
Human recombinant ER Resident 27 (from cells)
Protéines et Peptides
Endoplasmic reticulum resident protein 27, also known as ER protein 27, C12orf46 and ERP27, is an endoplasmic reticulum luminal protein which is a member of the protein disulfide isomerase family. ERP27 contains one thioredoxin domain and does not contain a CXXC active site motif. ERP27 is widely expressed in many tissues; it has highest expression in pancreas, with lower levels in spleen, lung, kidney, thymus, and bone marrow. ERP27 interacts with PDIA3 and binds somatostatin-14 via hydrophobic interactions. ERP27 may act as a protease, protein disulfide isomerase, thiol-disulfide oxidase or phospholipase.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité