Human recombinant proenkephalin-A (from cells)
Fournisseur: ProSci Inc.
91-769
PRSI91-769EA 852 EUR InStock
PRSI91-769
Human recombinant proenkephalin-A (from cells)
Protéines et Peptides
Proenkephalin-A is a secreted protein that belongs to the opioid neuropeptide precursor family. Proenkephalin-A is an endogenous opioid polypeptide hormone which, via proteolyic cleavage, produces the enkephalin peptides [Met]enkephalin, and to a lesser extent, [Leu]enkephalin. Met- and Leu-enkephalins compete with and mimic the effects of opiate drugs. They play a role in a number of physiologic functions, including pain perception and responses to stress. Proenkephalin-A (114-133) and Proenkephalin-A (237-258) increase glutamate release in the striatum. Proenkephalin-A (114-133) decreases GABA concentration in the striatum.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité