Human recombinant Brevican Core (from Cells)
Fournisseur: ProSci Inc.
91-772
PRSI91-772EA 654 EUR InStock
PRSI91-772
Human recombinant Brevican Core (from Cells)
Protéines et Peptides
Brevican Core Protein (BCAN) is a secreted protein that belongs to the aggrecan/versican proteoglycan family. BCAN contains one C-type lectin domain, one EGF-like domain, one Ig-like V-type domain, one Sushi (CCP/SCR) domain and two Link domains. BCAN may play a role in the terminally differentiating and the adult nervous system during postnatal development. BCAN could stabilize interactions between hyaluronan (HA) and brain proteoglycans.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité