Human recombinant neuroligin 4 (from cells)
Fournisseur: ProSci Inc.
91-775
PRSI91-775EA 654 EUR InStock
PRSI91-775
Human recombinant neuroligin 4 (from cells)
Protéines et Peptides
Neuroligin 4, X-Linked (NLGN4X) is a single-pass type I membrane protein that belongs to the type-B carboxylesterase/lipase family. NLGN4X is detected at higher levels in heart and at lower levels in the liver, skeletal muscle, and pancreas. NLGN4X is a putative neuronal cell surface protein involved in cell-cell-interactions. NLGN4X may act as splice site-specific ligands for beta -neurexins. It has been shown that NLGN4X is involved in the formation and remodeling of central nervous system synapses. NLGN4X also interacts with discs, large (Drosophila) homolog 4 (DLG4). Defects in NLGN4X have been associated with autism and Asperger syndrome.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité