Human recombinant proprotein convertase 9 (from cells)
Fournisseur: ProSci Inc.
91-785
PRSI91-785EA 584 EUR InStock
PRSI91-785
Human recombinant proprotein convertase 9 (from cells)
Enzymes
Recombinant Human Proprotein Convertase Subtilisin/Kexin Type 9/PCSK9 (D374Y) is a gain of function mutant of human PCSK9 protein. Human PCSK9 is a secretory subtilase belonging to the proteinase K subfamily. PCSK9 is synthesised as a soluble zymogen that undergoes autocatalytic intramolecular processing in the ER, the pro domain and mature chain are secreted together through noncovalent interactions. PCSK9 binds with low-density lipoprotein receptor (LDLR) and it plays a major regulatory role in cholesterol homeostasis. Inhibition of PCSK9 function by preventing PCSK9/LDLR interaction is currently being explored as a means of lowering cholesterol levels. PCSK9 also binds to apolipoprotein receptor 2 (ApoER2), and play a role in the neural development.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité